missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Methylpiperazino)benzaldehyde, 97%, Maybridge

Chemical Identifiers

CAS 27913-99-1
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD00452233
InChI Key PFODEVGLOVUVHS-UHFFFAOYSA-N
Synonym 1-4-formylphenyl-4-methylpiperazine
PubChem CID 736533
IUPAC Name 4-(4-methylpiperazin-1-yl)benzaldehyde
SMILES CN1CCN(CC1)C2=CC=C(C=C2)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC18704DA SDS View Documents Fisher Scientific Company
CC18704DA
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

27913-99-1
204.273
PFODEVGLOVUVHS-UHFFFAOYSA-N
736533
CN1CCN(CC1)C2=CC=C(C=C2)C=O
C12H16N2O
MFCD00452233
1-4-formylphenyl-4-methylpiperazine
4-(4-methylpiperazin-1-yl)benzaldehyde

Specifications

Amber glass bottle
27913-99-1
100.0
C12H16N2O
1-4-formylphenyl-4-methylpiperazine
CN1CCN(CC1)C2=CC=C(C=C2)C=O
204.273
204.27
4-(4-Methylpiperazino)benzaldehyde
1g
96.5
97%
MFCD00452233
PFODEVGLOVUVHS-UHFFFAOYSA-N
4-(4-methylpiperazin-1-yl)benzaldehyde
736533
97%