missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Maybridge

Manufacturer:  Fisher Scientific Company CC45616DA

 View more versions of this product

Catalog No. CC45616DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

215.3
97%
C13H17N3
4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile
Amber glass bottle
97%
1g
100.0
MFCD08060531
96.5
125743-63-7

Chemical Identifiers

125743-63-7
100.0
C13H17N3
MFCD08060531
97%
96.5
97%
215.3
4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger