missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Maybridge

Chemical Identifiers

CAS 125743-63-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H17N3
Formula Weight 215.3
MDL Number MFCD08060531
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC45616DA SDS Fisher Scientific Company
CC45616DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45616DE Fisher Scientific Company
CC45616DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

125743-63-7
100.0
C13H17N3
MFCD08060531
97%
96.5
97%
215.3
4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile

Specifications

1g
96.5
Amber glass bottle
C13H17N3
MFCD08060531
97%
125743-63-7
100.0
97%
215.3
4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger