Please login to your online account to display your discounted pricing

4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochlorideHemihydrate, 95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45651EA

 View more versions of this product

Catalog No. CC45651EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 316.23
Quantity 10g
Assay 95%
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride 0.5 hydrate
Molecular Formula 0·5 H2O
Packaging Amber Glass Bottle
Assay Percent Range 95%
CAS 106261-49-8
Health Hazard 1 Warning

Specifications

Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride 0.5 hydrate
Assay 95%
Assay Percent Range 95%
Molecular Formula 0·5 H2O
Formula Weight 316.23
Accessories

Warning