missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methylphenyl)-3-thiosemicarbazide, 98+%

Manufacturer:  Alfa Aesar™ L1126814

 View more versions of this product

Catalog No. AAL1126814



Specifications

Specifications

4-(4-Methylphenyl)-3-thiosemicarbazide
25g
C8H11N3S
UN2811
4-4-methylphenyl-3-thiosemicarbazide, n-p-tolyl hydrazinecarbothioamide, n-4-methylphenyl hydrazinecarbothioamide, 3-amino-1-4-methylphenyl thiourea, 1-amino-3-4-methylphenyl thiourea, n1-4-methylphenyl hydrazine-1-carbothioamide, hydrazino 4-methylphenyl amino methane-1-thione, acmc-20amvq, 4-p-tolylthiosemicarbazide, 4-p-tolylthiosemicarbazid
CC1=CC=C(C=C1)NC(=S)NN
181.257
181.26
135°C to 136°C
13278-67-6
MFCD00041302
879741
IEAWRKQVDLFINI-UHFFFAOYSA-N
1-amino-3-(4-methylphenyl)thiourea
737132
≥98%

Chemical Identifiers

13278-67-6
181.257
IEAWRKQVDLFINI-UHFFFAOYSA-N
737132
CC1=CC=C(C=C1)NC(=S)NN
C8H11N3S
MFCD00041302
4-4-methylphenyl-3-thiosemicarbazide, n-p-tolyl hydrazinecarbothioamide, n-4-methylphenyl hydrazinecarbothioamide, 3-amino-1-4-methylphenyl thiourea, 1-amino-3-4-methylphenyl thiourea, n1-4-methylphenyl hydrazine-1-carbothioamide, hydrazino 4-methylphenyl amino methane-1-thione, acmc-20amvq, 4-p-tolylthiosemicarbazide, 4-p-tolylthiosemicarbazid
1-amino-3-(4-methylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents