missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Methylperhydro-1,4-diazepin-1-yl)benzylamine, 97%, Maybridge

Chemical Identifiers

CAS 448934-01-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H21N3
Formula Weight 219.33
MDL Number MFCD08690316
Chemical Name or Material 4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC60313DA SDS Fisher Scientific Company
CC60313DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60313CB SDS Fisher Scientific Company
CC60313CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

448934-01-8
100.0
C13H21N3
MFCD08690316
97%
96.5
97%
219.33
4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine

Specifications

219.33
100.0
96.5
Amber Glass Bottle
C13H21N3
97%
1g
MFCD08690316
448934-01-8
97%
4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.