Learn More
4,4'-Methylenebis(N,N-dimethylaniline), 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126740050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4, 4'-Methylenebis(N, N-dimethylaniline) | |
101-61-1 | |
100.0 | |
Beige-Brown or Gray or White | |
178°C | |
98% | |
C17H22N2 | |
MFCD00008317 | |
13, 239 | |
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone | |
JNRLEMMIVRBKJE-UHFFFAOYSA-N | |
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline | |
7567 | |
254.37 | |
Crystalline Powder, Crystals or Chunks |
99+% | |
99.0 | |
86.0°C to 90.0°C | |
390.0°C | |
Authentic | |
Glass bottle | |
H2NC6H4CH2C6H4NH2 | |
5 g | |
15, 6255 | |
Solubility in water: insoluble. Other solubilities: soluble in benzene,ether,carbon disulfide and,acids,slightly soluble in cold alcohol,more soluble in,hot alcohol | |
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C | |
254.37 | |
CHEBI:34370 | |
98% |
Chemical Identifiers
101-61-1 | |
254.37 | |
JNRLEMMIVRBKJE-UHFFFAOYSA-N | |
7567 | |
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline |
C17H22N2 | |
MFCD00008317 | |
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone | |
CHEBI:34370 | |
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C |
Safety and Handling
GHS H Statement:
May cause cancer.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-959-2
RTECSNumber : BY5250000
TSCA : TSCA
RUO – Research Use Only