Learn More
4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 450240010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4, 4'-Methylenebis(2-methylcyclohexylamine) | |
0.9400g/mL | |
113°C | |
Glass bottle | |
MFCD00075535 | |
0.94 | |
IGSBHTZEJMPDSZ-UHFFFAOYNA-N | |
4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine | |
91555 | |
238.41 |
6864-37-5 | |
93.0°C to 100.0°C | |
98.5% min. (GC) | |
C15H30N2 | |
1L | |
4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine | |
CC1CC(CC2CCC(N)C(C)C2)CCC1N | |
238.42 | |
142 mPa.s (20°C) | |
99% |
Chemical Identifiers
6864-37-5 | |
238.42 | |
IGSBHTZEJMPDSZ-UHFFFAOYNA-N | |
91555 | |
CC1CC(CC2CCC(N)C(C)C2)CCC1N |
C15H30N2 | |
MFCD00075535 | |
4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine | |
4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 229-962-1
RUO â Research Use Only