missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(4-Methyl-2-piperidinylsulfonyl)acetanilide, 97%

Chemical Identifiers

CAS 1858240-55-7
Molecular Formula C14H20N2O3S
Molecular Weight (g/mol) 296.385
MDL Number MFCD22683128
InChI Key LUPVXSOGPVCCJN-UHFFFAOYSA-N
Synonym n-4-4-methylpiperidin-2-ylsulfonyl phenyl acetamide, 4'-4-methyl-2-piperidinylsulfonyl acetanilide
PubChem CID 121237459
IUPAC Name N-[4-(4-methylpiperidin-2-yl)sulfonylphenyl]acetamide
SMILES CC1CCNC(C1)S(=O)(=O)C2=CC=C(C=C2)NC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60478MD View Documents Alfa Aesar™
H60478MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6047803 View Documents Alfa Aesar™
H6047803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1858240-55-7
296.385
LUPVXSOGPVCCJN-UHFFFAOYSA-N
121237459
CC1CCNC(C1)S(=O)(=O)C2=CC=C(C=C2)NC(=O)C
C14H20N2O3S
MFCD22683128
n-4-4-methylpiperidin-2-ylsulfonyl phenyl acetamide, 4'-4-methyl-2-piperidinylsulfonyl acetanilide
N-[4-(4-methylpiperidin-2-yl)sulfonylphenyl]acetamide

Specifications

250mg
C14H20N2O3S
n-4-4-methylpiperidin-2-ylsulfonyl phenyl acetamide, 4'-4-methyl-2-piperidinylsulfonyl acetanilide
CC1CCNC(C1)S(=O)(=O)C2=CC=C(C=C2)NC(=O)C
296.385
296.38
4'-(4-Methyl-2-piperidinylsulfonyl)acetanilide
1858240-55-7
MFCD22683128
LUPVXSOGPVCCJN-UHFFFAOYSA-N
N-[4-(4-methylpiperidin-2-yl)sulfonylphenyl]acetamide
121237459
97%