missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methyl-1-piperazinylmethyl)benzoic acid, 97%

Manufacturer:  Alfa Aesar™ H5412003

 View more versions of this product

Catalog No. AAH5412003



Description

Description

Applications
4-(4-Methyl-1-piperazinylmethyl)benzoic acid is used as Imatinib intermediate, as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, acids, bases.
Specifications

Specifications

4-(4-Methyl-1-piperazinylmethyl)benzoic acid
106261-48-7
MFCD01632530
Slightly soluble in water.
CN1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
234.299
234.3
1g
C13H18N2O2
4-4-methyl-1-piperazinylmethyl benzoic acid
ZJUXJQSYXBYFFO-UHFFFAOYSA-N
4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
4769782
97%

Chemical Identifiers

106261-48-7
234.299
ZJUXJQSYXBYFFO-UHFFFAOYSA-N
4769782
CN1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
C13H18N2O2
MFCD01632530
4-4-methyl-1-piperazinylmethyl benzoic acid
4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents