Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%

$183.18 - $535.68

Chemical Identifiers

Chemical Name or Material 4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
CAS 938043-30-2
Molecular Formula C18H29BN2O2
Formula Weight 316.25
MDL Number MFCD09266182
Synonym 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH2849503

 
alfa aesar™
H2849503
Each for $183.18

AAH2849506

 
alfa aesar™
H2849506
Each for $535.68
Description & Specifications

Specifications

Formula Weight 316.25
Chemical Name or Material 4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
MDL Number MFCD09266182
Melting Point 61° to 65°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Molecular Formula C18H29BN2O2
CAS 938043-30-2
Synonym 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.