Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%

$88.59 - $257.66

Chemical Identifiers

Chemical Name or Material 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
CAS 832114-06-4
Molecular Formula C18H27BN2O3
Formula Weight 330.23
MDL Number MFCD05863914
Synonym 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH2821603

  SDS
alfa aesar™
H2821603
Each for $88.59

AAH2821606

  SDS
alfa aesar™
H2821606
Each for $257.66
Description & Specifications

Specifications

Formula Weight 330.23
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 832114-06-4
Chemical Name or Material 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C18H27BN2O3
MDL Number MFCD05863914
Synonym 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Melting Point 110° to 113°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.