missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H53296MD

 View more versions of this product

Catalog No. AAH53296MD



Specifications

Specifications

4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride
250mg
C12H19BClN3O3
4-4-methylpiperazin-1-yl carbamoyl phenyl boronic acid hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl phenylboronic acid hydrochloride, c12h17bn2o3.clh, n-4-methylpiperazinyl 4-boronobenzamide hcl, n4-methylpiperazinyl 4-boronobenzamide hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl-phenyl boronic acid hydrochloride
299.56
95%
187°C to 189°C
913835-43-5
MFCD09027217
DPTCRDJSYQTDGH-UHFFFAOYSA-N
284.55

Chemical Identifiers

913835-43-5
299.56
DPTCRDJSYQTDGH-UHFFFAOYSA-N
C12H19BClN3O3
MFCD09027217
4-4-methylpiperazin-1-yl carbamoyl phenyl boronic acid hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl phenylboronic acid hydrochloride, c12h17bn2o3.clh, n-4-methylpiperazinyl 4-boronobenzamide hcl, n4-methylpiperazinyl 4-boronobenzamide hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl-phenyl boronic acid hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents