missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester

$36.64 - $568.00

Chemical Identifiers

CAS 871366-38-0
Molecular Formula C20H24BNO3
Formula Weight 337.22
MDL Number MFCD10567027
Synonym 1-Methoxyphenyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Chemical Name or Material 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5111603 SDS Alfa Aesar™
H5111603
1g Each for $36.64
AAH5111606 SDS Alfa Aesar™
H5111606
5g Each for $139.76
AAH5111614 SDS Alfa Aesar™
H5111614
25g Each for $568.00
Specifications Description & Specifications

Chemical Identifiers

871366-38-0
337.22
1-Methoxyphenyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
C20H24BNO3
MFCD10567027
4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester

Specifications

871366-38-0
337.22
1-Methoxyphenyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester
C20H24BNO3
MFCD10567027
1g