missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 4-(4-Methoxyphenylethynyl)benzeneboronic acid pinacol ester,95%
$62.89 - $246.20
Chemical Identifiers
CAS | 1190376-24-9 |
---|---|
Molecular Formula | C21H23BO3 |
Molecular Weight (g/mol) | 334.222 |
MDL Number | MFCD16294533 |
InChI Key | RRYKLXOOBFDYDY-UHFFFAOYSA-N |
Synonym | 2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, amtb224 |
PubChem CID | 71306524 |
IUPAC Name | 2-[4-[2-(4-methoxyphenyl)ethynyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CC3=CC=C(C=C3)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5191403
|
Alfa Aesar™
H5191403 |
1g |
Each for $62.89
|
|
AAH5191406
|
Alfa Aesar™
H5191406 |
5g |
Each for $246.20
|
|
Chemical Identifiers
1190376-24-9 | |
334.222 | |
RRYKLXOOBFDYDY-UHFFFAOYSA-N | |
71306524 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CC3=CC=C(C=C3)OC |
C21H23BO3 | |
MFCD16294533 | |
2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, amtb224 | |
2-[4-[2-(4-methoxyphenyl)ethynyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
1g | |
C21H23BO3 | |
2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, amtb224 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CC3=CC=C(C=C3)OC | |
334.222 | |
334.22 | |
4-(4-Methoxyphenylethynyl)benzeneboronic acid pinacol ester |
1190376-24-9 | |
MFCD16294533 | |
RRYKLXOOBFDYDY-UHFFFAOYSA-N | |
2-[4-[2-(4-methoxyphenyl)ethynyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
71306524 | |
95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures