Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)pyrimidine-2-thiol, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(4-Methoxyphenyl)pyrimidine-2-thiol
CAS 175202-77-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H10N2OS
Formula Weight 218.28
MDL Number MFCD00085109
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM04163DA

  SDS
fisher scientific company
KM04163DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.28
Quantity 1g
Assay 95%
CAS Min % 95.0
Molecular Formula C11H10N2OS
Packaging Amber glass bottle
Chemical Name or Material 4-(4-Methoxyphenyl)pyrimidine-2-thiol
MDL Number MFCD00085109
CAS 175202-77-4
Assay Percent Range 95%