Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)pyrimidine-2-thiol, ≥95%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  KM04163DA

 View more versions of this product

Catalog No. KM04163DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 218.28
Quantity 1g
Assay 95%
CAS Min % 95.0
Molecular Formula C11H10N2OS
Packaging Amber glass bottle
Chemical Name or Material 4-(4-Methoxyphenyl)pyrimidine-2-thiol
CAS 175202-77-4
MDL Number MFCD00085109
Assay Percent Range 95%

Specifications

Chemical Name or Material 4-(4-Methoxyphenyl)pyrimidine-2-thiol
CAS 175202-77-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H10N2OS
Formula Weight 218.28
MDL Number MFCD00085109