Learn More
4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals
$137.54 - $458.23
Chemical Identifiers
CAS | 4521-28-2 |
---|---|
Molecular Formula | C11H14O3 |
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD00004404 |
InChI Key | LZHMNCJMXQKSBY-UHFFFAOYSA-N |
Synonym | 4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid |
PubChem CID | 78280 |
IUPAC Name | 4-(4-methoxyphenyl)butanoic acid |
SMILES | COC1=CC=C(C=C1)CCCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC172440050
|
Thermo Scientific Chemicals
172440050 |
5 g | Glass bottle |
Each for $137.54
|
|
||||
AC172440250
|
Thermo Scientific Chemicals
172440250 |
25 g | Glass bottle |
Each for $458.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4521-28-2 | |
194.23 | |
LZHMNCJMXQKSBY-UHFFFAOYSA-N | |
78280 | |
COC1=CC=C(C=C1)CCCC(=O)O |
C11H14O3 | |
MFCD00004404 | |
4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid | |
4-(4-methoxyphenyl)butanoic acid |
Specifications
4521-28-2 | |
100.0 | |
Cream to Yellow | |
98% | |
C11H14O3 | |
MFCD00004404 | |
10,IV,683 | |
LZHMNCJMXQKSBY-UHFFFAOYSA-N | |
4-(4-methoxyphenyl)butanoic acid | |
78280 | |
98% | |
4-(4-Methoxyphenyl)butyric acid |
97.5 | |
56.0°C to 61.0°C | |
Authentic | |
Glass bottle | |
CH3OC6H4(CH2)3CO2H | |
5 g | |
4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid | |
COC1=CC=C(C=C1)CCCC(=O)O | |
194.23 | |
194.23 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 224-849-3
RUO – Research Use Only