Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15NOS
Formula Weight 245.34
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW01484DA

 
fisher scientific company
SEW01484DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.34
Quantity 1g
Assay 97%
Chemical Name or Material 4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Assay Percent Range 97%
Molecular Formula C14H15NOS
CAS 213462-19-2
Packaging Amber Glass Bottle