Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW01484DA

 View more versions of this product

Catalog No. SEW01484DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 245.34
Quantity 1g
Assay 97%
Chemical Name or Material 4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
CAS 213462-19-2
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H15NOS

Specifications

Chemical Name or Material 4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15NOS
Formula Weight 245.34