missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methoxyphenyl)-2-methylthiazole, 97%

$79.42 - $1,261.46

Chemical Identifiers

CAS 50834-78-1
Molecular Formula C11H11NOS
Molecular Weight (g/mol) 205.275
MDL Number MFCD00460502
InChI Key OVKCVKLSCKKRJY-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl-2-methylthiazole, 4-4-methoxy-phenyl-2-methyl-thiazole, 4-4-methoxyphenyl-2-methyl-1,3-thiazole, thiazole, 4-4-methoxyphenyl-2-methyl, acmc-1ampl, 2-methyl-4-4-methoxyphenyl thiazole, 4-methoxy-1-2-methyl 1,3-thiazol-4-yl benzene
PubChem CID 768765
IUPAC Name 4-(4-methoxyphenyl)-2-methyl-1,3-thiazole
SMILES CC1=NC(=CS1)C2=CC=C(C=C2)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6606803 View Documents Alfa Aesar™
H6606803
1g Each for $79.42
 
AAH6606806 View Documents Alfa Aesar™
H6606806
5g Each for $303.80
 
AAH6606814 View Documents Alfa Aesar™
H6606814
25g Each for $1,261.46
 
Specifications

Chemical Identifiers

50834-78-1
205.275
OVKCVKLSCKKRJY-UHFFFAOYSA-N
768765
CC1=NC(=CS1)C2=CC=C(C=C2)OC
C11H11NOS
MFCD00460502
4-4-methoxyphenyl-2-methylthiazole, 4-4-methoxy-phenyl-2-methyl-thiazole, 4-4-methoxyphenyl-2-methyl-1,3-thiazole, thiazole, 4-4-methoxyphenyl-2-methyl, acmc-1ampl, 2-methyl-4-4-methoxyphenyl thiazole, 4-methoxy-1-2-methyl 1,3-thiazol-4-yl benzene
4-(4-methoxyphenyl)-2-methyl-1,3-thiazole

Specifications

Cream
1g
C11H11NOS
4-4-methoxyphenyl-2-methylthiazole, 4-4-methoxy-phenyl-2-methyl-thiazole, 4-4-methoxyphenyl-2-methyl-1,3-thiazole, thiazole, 4-4-methoxyphenyl-2-methyl, acmc-1ampl, 2-methyl-4-4-methoxyphenyl thiazole, 4-methoxy-1-2-methyl 1,3-thiazol-4-yl benzene
CC1=NC(=CS1)C2=CC=C(C=C2)OC
205.275
205.28
97%
64°C to 69°C
50834-78-1
MFCD00460502
OVKCVKLSCKKRJY-UHFFFAOYSA-N
4-(4-methoxyphenyl)-2-methyl-1,3-thiazole
768765
Solid
4-(4-Methoxyphenyl)-2-methylthiazole