missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Methoxyphenyl)-2-butanone, 99%

$34.95 - $489.49

Chemical Identifiers

CAS 104-20-1
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00008791
Beilstein 1869592
Synonym p-Anisylacetone
Chemical Name or Material 4- (4-Methoxyphenyl)-2-butanone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1101109 SDS Alfa Aesar™
A1101109
10g Each for $34.95
AAA1101118 SDS Alfa Aesar™
A1101118
50g Each for $112.51
AAA1101130 SDS Alfa Aesar™
A1101130
250g Each for $489.49
Specifications

Description

Applications
4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

104-20-1
178.23
1869592
4- (4-Methoxyphenyl)-2-butanone
C11H14O2
MFCD00008791
p-Anisylacetone

Specifications

104-20-1
178.23
147° to 148°C (8mmHg)
7° to 9°C
10g
4- (4-Methoxyphenyl)-2-butanone
C11H14O2
MFCD00008791
1869592
p-Anisylacetone
>110°C (230°F)
99%

Safety and Handling

RTECSNumber : EL9085500

TSCA : TSCA