Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)-2-butanone, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  313980500

 View more versions of this product

Catalog No. AC313980500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 178.23
Quantity 50g
Color Clear colorless to pale yellow
EINECS Number 203-184-2
TSCA TSCA
Density 1.04g/mL
RTECS Number EL9085500
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
Synonym Anisylacetone
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 4-(4-Methoxyphenyl)-2-butanone
Name Note 98%
Assay Percent Range 98%
Molecular Formula C11H14O2
Linear Formula CH3OC6H4CHCHCOCH3
MDL Number MFCD00008791
CAS 104-20-1

Specifications

Chemical Name or Material 4-(4-Methoxyphenyl)-2-butanone
Name Note 98%
CAS 104-20-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H14O2
Linear Formula CH3OC6H4CHCHCOCH3
Formula Weight 178.23
MDL Number MFCD00008791
Synonym Anisylacetone