missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)-2-butanone, 98%, ACROS Organics™

$109.78 - $109.78

Chemical Identifiers

CAS 104-20-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H14O2
Linear Formula CH3OC6H4CHCHCOCH3
Formula Weight 178.23
MDL Number MFCD00008791
Synonym Anisylacetone
Chemical Name or Material 4-(4-Methoxyphenyl)-2-butanone, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC313980500 SDS Thermo Fisher Scientific
313980500
Glass bottle 50g Each for $109.78
AC313980050 SDS Thermo Fisher Scientific
313980050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

104-20-1
100.0
C11H14O2
178.23
Anisylacetone
≥97.5%
97.5
98%
CH3OC6H4CHCHCOCH3
MFCD00008791
4-(4-Methoxyphenyl)-2-butanone, 98%

Specifications

104-20-1
100.0
Authentic
Liquid
1.04g/mL
CH3OC6H4CHCHCOCH3
MFCD00008791
Anisylacetone
4-(4-Methoxyphenyl)-2-butanone, 98%
97.5
Glass bottle
1.5190 to 1.5210 (20°C, 589nm)
98%
C11H14O2
178.23
Clear colorless to pale yellow
50g
≥97.5%

Safety and Handling

EINECSNumber : 203-184-2

RTECSNumber : EL9085500

TSCA : TSCA