Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester, 95%

$112.51 - $444.85

Chemical Identifiers

Chemical Name or Material 4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester
CAS 1448637-05-5
Molecular Formula C22H23BO4
Formula Weight 362.23
MDL Number MFCD16294527
Synonym 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenylethynyl]benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190003

  SDS
alfa aesar™
H5190003
1g Each for $112.51

AAH5190006

  SDS
alfa aesar™
H5190006
5g Each for $444.85
Description & Specifications

Specifications

Formula Weight 362.23
Chemical Name or Material 4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester
Assay 95%
Molecular Formula C22H23BO4
MDL Number MFCD16294527
CAS 1448637-05-5
Synonym 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenylethynyl]benzoic acid methyl ester
Quantity 1g