missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445080050

 View more versions of this product

Catalog No. AC445080050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester
5g
96% min. (GC)
1-isopropyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-isopropyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-propan-2-yl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-isopropylpiperazin-1-yl phenylboronic acid pinacol ester, 4-4-isopropylpiperazinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl benzeneboronic acid, pinacol ester, 4-4-isopropylpiperizinyl phenylboronic acid pinacol ester, 4-4-isopropylpiperizinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl phenylboronicacid,pinacolester
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(C)C
330.28
330.28
139°C to 141°C
1073354-18-3
C19H31BN2O2
CSORKGLMGUQQOY-UHFFFAOYSA-N
1-propan-2-yl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
17750254
97%

Chemical Identifiers

1073354-18-3
330.28
1-isopropyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-isopropyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-propan-2-yl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-isopropylpiperazin-1-yl phenylboronic acid pinacol ester, 4-4-isopropylpiperazinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl benzeneboronic acid, pinacol ester, 4-4-isopropylpiperizinyl phenylboronic acid pinacol ester, 4-4-isopropylpiperizinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl phenylboronicacid,pinacolester
1-propan-2-yl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
C19H31BN2O2
CSORKGLMGUQQOY-UHFFFAOYSA-N
17750254
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(C)C
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents