missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Isopropylidenediphenoxyacetic Acid, TCI America™

$288.75 - $864.73

Chemical Identifiers

CAS 3539-42-2
Molecular Formula C19H20O6
Molecular Weight (g/mol) 344.363
MDL Number MFCD00060114
InChI Key ZYGPJSLWIIMUMO-UHFFFAOYSA-N
Synonym Bisphenol A O,O-Diacetic Acid
PubChem CID 12359775
IUPAC Name 2-[4-[2-[4-(carboxymethoxy)phenyl]propan-2-yl]phenoxy]acetic acid
SMILES CC(C)(C1=CC=C(C=C1)OCC(=O)O)C2=CC=C(C=C2)OCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I03565G View Documents TCI America
I03565G
5g Each for $288.75
 
I035625G View Documents TCI America
I035625G
25g Each for $864.73
 
Specifications

Chemical Identifiers

3539-42-2
344.363
ZYGPJSLWIIMUMO-UHFFFAOYSA-N
12359775
CC(C)(C1=CC=C(C=C1)OCC(=O)O)C2=CC=C(C=C2)OCC(=O)O
C19H20O6
MFCD00060114
Bisphenol A O,O-Diacetic Acid
2-[4-[2-[4-(carboxymethoxy)phenyl]propan-2-yl]phenoxy]acetic acid

Specifications

White-Yellow
5g
C19H20O6
Bisphenol A O,O-Diacetic Acid
CC(C)(C1=CC=C(C=C1)OCC(=O)O)C2=CC=C(C=C2)OCC(=O)O
344.363
344.36
≥98.0% (HPLC,T)
179°C
3539-42-2
MFCD00060114
ZYGPJSLWIIMUMO-UHFFFAOYSA-N
2-[4-[2-[4-(carboxymethoxy)phenyl]propan-2-yl]phenoxy]acetic acid
12359775
Crystal-Powder at 20°C
4,4′-Isopropylidenediphenoxyacetic Acid
Safety and Handling

Safety and Handling

TSCA : No