Please login to your online account to display your discounted pricing

4-(4-Iodophenyl)morpholine, Maybridge

$102.75 - $102.75

Chemical Identifiers

Chemical Name or Material 4-(4-Iodophenyl)morpholine
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12INO
Formula Weight 289.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC17411CB

  SDS
fisher scientific company
CC17411CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC17411DE

 
fisher scientific company
CC17411DE
Amber Glass Bottle 5g Each for $102.75
Description & Specifications

Specifications

Formula Weight 289.11
Quantity 250mg
Assay 97%
Chemical Name or Material 4-(4-Iodophenyl)morpholine
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 87350-77-4
Molecular Formula C10H12INO