Please login to your online account to display your discounted pricing

4-(4-Iodophenyl)morpholine, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC17411DE

 View more versions of this product

Catalog No. CC17411DE

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 289.11
Quantity 5g
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12INO
Packaging Amber Glass Bottle
Chemical Name or Material 4-(4-Iodophenyl)morpholine
CAS 87350-77-4

Specifications

Chemical Name or Material 4-(4-Iodophenyl)morpholine
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12INO
Formula Weight 289.11