missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Hydroxyphenyl)cyclohexanone, 97+%

Manufacturer:  Alfa Aesar™ H5576703

 View more versions of this product

Catalog No. AAH5576703



Specifications

Specifications

4-(4-Hydroxyphenyl)cyclohexanone
1g
C12H14O2
3269332
SLJYPZJZQIHNGU-UHFFFAOYSA-N
4-(4-hydroxyphenyl)cyclohexan-1-one
4598916
≥97%
170°C to 175°C
105640-07-1
MFCD00210693
4-4'-hydroxyphenyl-cyclohexanone
C1CC(=O)CCC1C2=CC=C(C=C2)O
190.242
190.24

Chemical Identifiers

105640-07-1
190.242
SLJYPZJZQIHNGU-UHFFFAOYSA-N
4598916
C1CC(=O)CCC1C2=CC=C(C=C2)O
C12H14O2
MFCD00210693
4-4'-hydroxyphenyl-cyclohexanone
4-(4-hydroxyphenyl)cyclohexan-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents