missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Hydroxyphenoxy)benzoic Acid, TCI America™

$212.36 - $678.34

Chemical Identifiers

CAS 500-76-5
Molecular Formula C13H10O4
Molecular Weight (g/mol) 230.219
MDL Number MFCD00059645
InChI Key KLXPCYHWTLAVLN-UHFFFAOYSA-N
PubChem CID 615542
IUPAC Name 4-(4-hydroxyphenoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H06425G View Documents TCI America
H06425G
5g Each for $678.34
 
H06421G View Documents TCI America
H06421G
1g Each for $212.36
 
Specifications

Chemical Identifiers

500-76-5
230.219
KLXPCYHWTLAVLN-UHFFFAOYSA-N
4-(4-hydroxyphenoxy)benzoic acid
C13H10O4
MFCD00059645
615542
C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)O

Specifications

Undesignated
5g
C13H10O4
KLXPCYHWTLAVLN-UHFFFAOYSA-N
4-(4-hydroxyphenoxy)benzoic acid
615542
Crystal-Powder at 20°C
4-(4-Hydroxyphenoxy)benzoic Acid
196°C
500-76-5
MFCD00059645
C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)O
230.219
230.22
≥99.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No