missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Hydroxybutyl)thiomorpholine 1,1-Dioxide, TCI America™

$242.92 - $847.94

Chemical Identifiers

CAS 59801-41-1
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
InChI Key STKAHXPYHDVHGO-UHFFFAOYSA-N
Synonym 4-(1,1-Dioxothiomorpholino)-1-butanol
PubChem CID 12324072
IUPAC Name 4-(1,1-dioxo-1,4-thiazinan-4-yl)butan-1-ol
SMILES C1CS(=O)(=O)CCN1CCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H11425G View Documents TCI America
H11425G
5g Each for $242.92
 
H114225G View Documents TCI America
H114225G
25g Each for $847.94
 
Specifications

Chemical Identifiers

59801-41-1
207.288
4-(1,1-Dioxothiomorpholino)-1-butanol
4-(1,1-dioxo-1,4-thiazinan-4-yl)butan-1-ol
C8H17NO3S
STKAHXPYHDVHGO-UHFFFAOYSA-N
12324072
C1CS(=O)(=O)CCN1CCCCO

Specifications

Undesignated
5g
C8H17NO3S
STKAHXPYHDVHGO-UHFFFAOYSA-N
4-(1,1-dioxo-1,4-thiazinan-4-yl)butan-1-ol
12324072
Crystal-Powder at 20°C
4-(4-Hydroxybutyl)thiomorpholine 1,1-Dioxide
65°C
59801-41-1
4-(1,1-Dioxothiomorpholino)-1-butanol
C1CS(=O)(=O)CCN1CCCCO
207.288
207.29
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator