missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Formylphenyl)morpholine, TCI America™

$38.19 - $320.84

Chemical Identifiers

CAS 1204-86-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
InChI Key FOAQOAXQMISINY-UHFFFAOYSA-N
Synonym 4-morpholinobenzaldehyde, 4-morpholin-4-yl-benzaldehyde, 4-4-morpholinyl benzaldehyde, 4-4-formylphenyl morpholine, 4-morpholin-4-yl benzaldehyde, 4-morpholinobenzenecarbaldehyde, benzaldehyde, 4-4-morpholinyl, akos bc-2976, acmc-1bqjc, 4-morpholinylbenzaldehyde
PubChem CID 291349
IUPAC Name 4-morpholin-4-ylbenzaldehyde
SMILES C1COCCN1C2=CC=C(C=C2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M16255G View Documents TCI America
M16255G
5g Each for $96.25
 
M16251G View Documents TCI America
M16251G
1g Each for $38.19
 
M162525G View Documents TCI America
M162525G
25g Each for $320.84
 
Specifications

Chemical Identifiers

1204-86-0
191.23
4-morpholinobenzaldehyde, 4-morpholin-4-yl-benzaldehyde, 4-4-morpholinyl benzaldehyde, 4-4-formylphenyl morpholine, 4-morpholin-4-yl benzaldehyde, 4-morpholinobenzenecarbaldehyde, benzaldehyde, 4-4-morpholinyl, akos bc-2976, acmc-1bqjc, 4-morpholinylbenzaldehyde
4-morpholin-4-ylbenzaldehyde
C11H13NO2
FOAQOAXQMISINY-UHFFFAOYSA-N
291349
C1COCCN1C2=CC=C(C=C2)C=O

Specifications

Undesignated
5g
C11H13NO2
FOAQOAXQMISINY-UHFFFAOYSA-N
4-morpholin-4-ylbenzaldehyde
291349
Crystal-Powder at 20°C
4-(4-Formylphenyl)morpholine
63°C
1204-86-0
4-morpholinobenzaldehyde, 4-morpholin-4-yl-benzaldehyde, 4-4-morpholinyl benzaldehyde, 4-4-formylphenyl morpholine, 4-morpholin-4-yl benzaldehyde, 4-morpholinobenzenecarbaldehyde, benzaldehyde, 4-4-morpholinyl, akos bc-2976, acmc-1bqjc, 4-morpholinylbenzaldehyde
C1COCCN1C2=CC=C(C=C2)C=O
191.23
191.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No