missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorophenylsulfamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54141MD

 View more versions of this product

Catalog No. AAH54141MD



Specifications

Specifications

4-(4-Fluorophenylsulfamoyl)benzeneboronic acid
957121-13-0
MFCD09800889
4-4-fluorophenyl sulfamoyl phenyl boronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)F)(O)O
295.091
295.09
250mg
C12H11BFNO4S
23848264
MFWQIJUPTCPQPU-UHFFFAOYSA-N
[4-[(4-fluorophenyl)sulfamoyl]phenyl]boronic acid
44717721
97%

Chemical Identifiers

957121-13-0
295.091
MFWQIJUPTCPQPU-UHFFFAOYSA-N
44717721
B(C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)F)(O)O
C12H11BFNO4S
MFCD09800889
4-4-fluorophenyl sulfamoyl phenyl boronic acid
[4-[(4-fluorophenyl)sulfamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents