Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorophenyl)butyric acid, 97%

$91.73 - $232.43

Chemical Identifiers

Chemical Name or Material 4-(4-Fluorophenyl)butyric acid
CAS 589-06-0
Molecular Formula C10H11FO2
Formula Weight 182.2
MDL Number MFCD03788503
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603503

  SDS
alfa aesar™
H2603503
1g Each for $232.43

AAH26035MD

  SDS
alfa aesar™
H26035MD
250mg Each for $91.73
Description & Specifications

Specifications

Formula Weight 182.2
Chemical Name or Material 4-(4-Fluorophenyl)butyric acid
Assay 97%
Molecular Formula C10H11FO2
MDL Number MFCD03788503
CAS 589-06-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 44° to 46°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.