Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorophenyl)-3-thiosemicarbazide, 98%

Chemical Identifiers

Chemical Name or Material 4-(4-Fluorophenyl)-3-thiosemicarbazide
CAS 330-94-9
Molecular Formula C7H8FN3S
Formula Weight 185.22
MDL Number MFCD00041294
UN Number UN2811
Beilstein 2804705
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0948206

 
alfa aesar™
L0948206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.22
CAS 330-94-9
Chemical Name or Material 4-(4-Fluorophenyl)-3-thiosemicarbazide
Assay 98%
Molecular Formula C7H8FN3S
MDL Number MFCD00041294
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Odor Odorless
Beilstein 2804705
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point ≈179°C
Quantity 5g

Applications
It is an important pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.