Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorophenoxy)benzylamine hydrochloride, 97%

$53.73 - $635.72

Chemical Identifiers

Chemical Name or Material 4-(4-Fluorophenoxy)benzylamine hydrochloride
CAS 568565-86-6
Molecular Formula C13H12FNO·HCl
Formula Weight 253.7
MDL Number MFCD01862521
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2018103

  SDS
alfa aesar™
L2018103
1g Each for $53.73

AAL2018106

  SDS
alfa aesar™
L2018106
5g Each for $166.24

AAL2018114

  SDS
alfa aesar™
L2018114
25g Each for $635.72
Description & Specifications

Specifications

Formula Weight 253.7
Chemical Name or Material 4-(4-Fluorophenoxy)benzylamine hydrochloride
CAS 568565-86-6
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C13H12FNO·HCl
MDL Number MFCD01862521
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 238° to 240°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.