missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Fluorobenzyloxy)benzaldehyde, 97%
CAS: 56442-17-2 | C14H11FO2 | 230.24 g/mol
$77.32 - $257.74
Chemical Identifiers
| CAS | 56442-17-2 |
|---|---|
| Molecular Formula | C14H11FO2 |
| Molecular Weight (g/mol) | 230.24 |
| MDL Number | MFCD00173650 |
| InChI Key | QGULWBQOCMQNFD-UHFFFAOYSA-N |
| Synonym | 4-4-fluorobenzyloxy benzaldehyde, 4-4-fluorobenzyl oxy benzaldehyde, 4-4-fluorophenyl methoxy benzaldehyde, acmc-1b0gw, cbmicro_008348, 4-4-fluorobenzyl-oxy benzaldehyde, 4-4-fluoro-benzyloxy-benzaldehyde, benzaldehyde,4-4-fluorophenyl methoxy |
| PubChem CID | 735860 |
| IUPAC Name | 4-[(4-fluorophenyl)methoxy]benzaldehyde |
| SMILES | FC1=CC=C(COC2=CC=C(C=O)C=C2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1765706
|
Thermo Scientific Chemicals
A1765706 |
5 g |
Each for $77.32
|
|
|||||
|
AAA1765714
|
Thermo Scientific Chemicals
A1765714 |
25 g |
Each for $257.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56442-17-2 | |
| 230.24 | |
| QGULWBQOCMQNFD-UHFFFAOYSA-N | |
| 735860 | |
| FC1=CC=C(COC2=CC=C(C=O)C=C2)C=C1 |
| C14H11FO2 | |
| MFCD00173650 | |
| 4-4-fluorobenzyloxy benzaldehyde, 4-4-fluorobenzyl oxy benzaldehyde, 4-4-fluorophenyl methoxy benzaldehyde, acmc-1b0gw, cbmicro_008348, 4-4-fluorobenzyl-oxy benzaldehyde, 4-4-fluoro-benzyloxy-benzaldehyde, benzaldehyde,4-4-fluorophenyl methoxy | |
| 4-[(4-fluorophenyl)methoxy]benzaldehyde |
Specifications
| 56442-17-2 | |
| MFCD00173650 | |
| Air Sensitive | |
| QGULWBQOCMQNFD-UHFFFAOYSA-N | |
| 4-[(4-fluorophenyl)methoxy]benzaldehyde | |
| 735860 | |
| 97% |
| C14H11FO2 | |
| 5 g | |
| 4-4-fluorobenzyloxy benzaldehyde, 4-4-fluorobenzyl oxy benzaldehyde, 4-4-fluorophenyl methoxy benzaldehyde, acmc-1b0gw, cbmicro_008348, 4-4-fluorobenzyl-oxy benzaldehyde, 4-4-fluoro-benzyloxy-benzaldehyde, benzaldehyde,4-4-fluorophenyl methoxy | |
| FC1=CC=C(COC2=CC=C(C=O)C=C2)C=C1 | |
| 230.24 | |
| 230.24 | |
| 4-(4-Fluorobenzyloxy)benzaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only