missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorobenzyl)piperidine hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H6386014

 View more versions of this product

Catalog No. AAH6386014



Description

Description

Applications
Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Specifications

4-(4-Fluorobenzyl)piperidine hydrochloride
193357-52-7
MFCD03840140
OJKWWANXBGLGQN-UHFFFAOYSA-N
4-[(4-fluorophenyl)methyl]piperidine;hydrochloride
17039492
Solid
25g
C12H17ClFN
4-4-fluorobenzyl piperidine hydrochloride, 4-4-fluorobenzyl piperidine hcl, 4-4-fluoro-benzyl-piperidine hydrochloride, 4-4'-fluorobenzyl piperidine hydrochloride, piperidine, 4-4-fluorophenyl methyl-, hydrochloride, 4-4-fluorophenyl methyl piperidine hydrochloride, 4-4-fluorobenzyl piperidine hydrochloride salt, 4-4-fluorobenzyl piperidine hydrobromide, 4-4-fluorobenzyl piperidinehydrochloride, 4-4-fluorobenzyl-piperidine hydrochloride
C1CNCCC1CC2=CC=C(C=C2)F.Cl
229.723
229.72
97%

Chemical Identifiers

193357-52-7
229.723
OJKWWANXBGLGQN-UHFFFAOYSA-N
17039492
C1CNCCC1CC2=CC=C(C=C2)F.Cl
C12H17ClFN
MFCD03840140
4-4-fluorobenzyl piperidine hydrochloride, 4-4-fluorobenzyl piperidine hcl, 4-4-fluoro-benzyl-piperidine hydrochloride, 4-4'-fluorobenzyl piperidine hydrochloride, piperidine, 4-4-fluorophenyl methyl-, hydrochloride, 4-4-fluorophenyl methyl piperidine hydrochloride, 4-4-fluorobenzyl piperidine hydrochloride salt, 4-4-fluorobenzyl piperidine hydrobromide, 4-4-fluorobenzyl piperidinehydrochloride, 4-4-fluorobenzyl-piperidine hydrochloride
4-[(4-fluorophenyl)methyl]piperidine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents