Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%

$93.43 - $332.49

Chemical Identifiers

Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride
CAS 25519-78-2
Molecular Formula C12H14FNO·HCl
Formula Weight 243.71
MDL Number MFCD00044912
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2314203

 
alfa aesar™
B2314203
1g Each for $93.43

AAB2314206

 
alfa aesar™
B2314206
5g Each for $332.49
Description & Specifications

Specifications

Formula Weight 243.71
Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride
Assay 98%
Molecular Formula C12H14FNO·HCl
MDL Number MFCD00044912
CAS 25519-78-2
Quantity 1g