Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluorobenzoyl)butyric acid, 98%

$397.78 - $397.78

Chemical Identifiers

Chemical Name or Material 4-(4-Fluorobenzoyl)butyric acid
CAS 149437-76-3
Molecular Formula C11H11FO3
Formula Weight 210.21
MDL Number MFCD00667222
Synonym 5-(4-Fluorophenyl)-5-oxopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2583906

  SDS
alfa aesar™
H2583906
5g Each for $397.78
Description & Specifications

Specifications

Formula Weight 210.21
CAS 149437-76-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C11H11FO3
Synonym 5-(4-Fluorophenyl)-5-oxopentanoic acid
Health Hazard 1 Warning
Assay 98%
MDL Number MFCD00667222
Chemical Name or Material 4-(4-Fluorobenzoyl)butyric acid
Melting Point 141° to 144°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.