Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluoro-2-nitrophenyl)morpholine, 97%

Chemical Identifiers

Chemical Name or Material 4-(4-Fluoro-2-nitrophenyl)morpholine
CAS 238418-75-2
Formula Weight 226.2
MDL Number MFCD00798369
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6324703

 
alfa aesar™
H6324703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6324706

 
alfa aesar™
H6324706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6324714

 
alfa aesar™
H6324714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.2
Chemical Name or Material 4-(4-Fluoro-2-nitrophenyl)morpholine
Physical Form Solid
Assay 97%
MDL Number MFCD00798369
CAS 238418-75-2
Quantity 1g