missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Fluoro-2-nitrophenyl)morpholine, 97%

Manufacturer:  Alfa Aesar™ H6324706

 View more versions of this product

Catalog No. AAH6324706



Specifications

Specifications

4-(4-Fluoro-2-nitrophenyl)morpholine
238418-75-2
MFCD00798369
XEVFYVWDTJTEHQ-UHFFFAOYSA-N
4-(4-fluoro-2-nitrophenyl)morpholine
2737431
Solid
5g
C10H11FN2O3
4-4-fluoro-2-nitrophenyl morpholine, acmc-1cbj8, 5-fluoro-2-morpholinonitrobenzene, 4-4-fluoro-2-nitro-phenyl-morpholine, morpholine,4-4-fluoro-2-nitrophenyl
C1COCCN1C2=C(C=C(C=C2)F)[N+](=O)[O-]
226.207
226.2
97%

Chemical Identifiers

238418-75-2
226.207
XEVFYVWDTJTEHQ-UHFFFAOYSA-N
2737431
C1COCCN1C2=C(C=C(C=C2)F)[N+](=O)[O-]
C10H11FN2O3
MFCD00798369
4-4-fluoro-2-nitrophenyl morpholine, acmc-1cbj8, 5-fluoro-2-morpholinonitrobenzene, 4-4-fluoro-2-nitro-phenyl-morpholine, morpholine,4-4-fluoro-2-nitrophenyl
4-(4-fluoro-2-nitrophenyl)morpholine
Documents