Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Ethylphenyl)-3-thiosemicarbazide, 98+%

Chemical Identifiers

Chemical Name or Material 4-(4-Ethylphenyl)-3-thiosemicarbazide
CAS 93693-01-7
Molecular Formula C9H13N3S
Formula Weight 195.29
MDL Number MFCD00041303
UN Number UN2811
Beilstein 2804749
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1236103

  SDS
alfa aesar™
L1236103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1236106

  SDS
alfa aesar™
L1236106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.29
CAS 93693-01-7
MDL Number MFCD00041303
Chemical Name or Material 4-(4-Ethylphenyl)-3-thiosemicarbazide
Molecular Formula C9H13N3S
Health Hazard 3 GHS P Statement
P280f-P307+P311
Wear protective clothing.
IF exposed: Call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Melting Point 128° to 131°C
Quantity 1g
UN Number UN2811
Assay ≥98%
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Beilstein 2804749
Safety and Handling

Danger

GHS P Statement
P280f-P307+P311
Wear protective clothing.
IF exposed: Call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.