missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Ethylphenyl)-3-thiosemicarbazide, 98+%

Manufacturer:  Alfa Aesar™ L1236103

 View more versions of this product

Catalog No. AAL1236103



Specifications

Specifications

4-(4-Ethylphenyl)-3-thiosemicarbazide
1g
C9H13N3S
UN2811
4-4-ethylphenyl-3-thiosemicarbazide, n-4-ethylphenyl hydrazinecarbothioamide, 3-amino-1-4-ethylphenyl thiourea, 4-ethylphenyl amino hydrazinomethane-1-thione, 1-amino-3-4-ethylphenyl thiourea, 4-4'-ethylphenyl-3-thiosemicarbazide
CCC1=CC=C(C=C1)NC(=S)NN
195.284
195.29
128°C to 131°C
93693-01-7
MFCD00041303
2804749
MXVVZBDSLDIVAW-UHFFFAOYSA-N
1-amino-3-(4-ethylphenyl)thiourea
3259670
≥98%

Chemical Identifiers

93693-01-7
195.284
MXVVZBDSLDIVAW-UHFFFAOYSA-N
3259670
CCC1=CC=C(C=C1)NC(=S)NN
C9H13N3S
MFCD00041303
4-4-ethylphenyl-3-thiosemicarbazide, n-4-ethylphenyl hydrazinecarbothioamide, 3-amino-1-4-ethylphenyl thiourea, 4-ethylphenyl amino hydrazinomethane-1-thione, 1-amino-3-4-ethylphenyl thiourea, 4-4'-ethylphenyl-3-thiosemicarbazide
1-amino-3-(4-ethylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P307+P311
Wear protective clothing.
IF exposed: Call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents