missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Ethylenedianiline, TCI America™

Manufacturer:  TCI America E03465G

 View more versions of this product

Catalog No. E03465G



Specifications

Specifications

4,4′-Ethylenedianiline
134°C
621-95-4
MFCD00007923
UHNUHZHQLCGZDA-UHFFFAOYSA-N
4-[2-(4-aminophenyl)ethyl]aniline
12138
Crystal-Powder at 20°C
Undesignated
5g
C14H16N2
4,4'-ethylenedianiline, 4,4'-diaminobibenzyl, 4-2-4-aminophenyl ethyl aniline, 4,4'-diaminodibenzyl, 1,2-bis p-aminophenyl ethane, benzenamine, 4,4'-1,2-ethanediyl bis, 4,4'-ethylenebis aniline, aniline, 4,4'-ethylenedi, .alpha.,.alpha.'-bi-p-toluidine, 4-2-4-aminophenyl ethyl phenylamine
C1=CC(=CC=C1CCC2=CC=C(C=C2)N)N
212.296
212.30
≥97.0% (GC,T)

Chemical Identifiers

621-95-4
212.296
UHNUHZHQLCGZDA-UHFFFAOYSA-N
12138
C1=CC(=CC=C1CCC2=CC=C(C=C2)N)N
C14H16N2
MFCD00007923
4,4'-ethylenedianiline, 4,4'-diaminobibenzyl, 4-2-4-aminophenyl ethyl aniline, 4,4'-diaminodibenzyl, 1,2-bis p-aminophenyl ethane, benzenamine, 4,4'-1,2-ethanediyl bis, 4,4'-ethylenebis aniline, aniline, 4,4'-ethylenedi, .alpha.,.alpha.'-bi-p-toluidine, 4-2-4-aminophenyl ethyl phenylamine
4-[2-(4-aminophenyl)ethyl]aniline
Safety and Handling

Safety and Handling

RTECSNumber : BY0200000

TSCA : Yes

Documents