Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Dipyridyl disulfide, 98%

$52.05 - $166.93

Chemical Identifiers

Chemical Name or Material 4,4'-Dipyridyl disulfide
CAS 2645-22-9
Molecular Formula C10H8N2S2
Formula Weight 220.32g/mol
MDL Number MFCD00006423
Beilstein 154742
Synonym 4,4'-Dithiodipyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2402303

 
alfa aesar™
B24023-03
1g Each for $52.05

AAB2402306

 
alfa aesar™
B24023-06
5g Each for $166.93
Description & Specifications

Specifications

Melting Point 74° to 78°C
Quantity 1g
Chemical Name or Material 4,4'-Dipyridyl disulfide
CAS 2645-22-9
Assay 98%
Molecular Formula C10H8N2S2
Formula Weight 220.32g/mol
MDL Number MFCD00006423
Beilstein 154742
Synonym 4,4'-Dithiodipyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C