missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Dipyridyl, 98%, anhydrous, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 276200250
Specifications
4, 4'-Dipyridyl | |
Glass bottle | |
1% Max. (K.F.) | |
White to Beige | |
25g | |
98% | |
(C5H4N)-(C5H4N) | |
16,25 | |
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridine, 4,4'-dipyridyl, 4,4-bipyridyl, 4,4-dipyridyl, 4-4-pyridyl pyridine, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb | |
MWVTWFVJZLCBMC-UHFFFAOYSA-N | |
4-pyridin-4-ylpyridine | |
11107 | |
156.19 | |
98% |
Authentic | |
0.1% max. | |
305.0°C | |
110.0°C to 114.0°C | |
553-26-4 | |
C10H8N2 | |
23,2 | |
15,3389 | |
Solubility in water: 4,5g/L (25°C). Other solubilities: freely soluble in alcohol, benzene, chloroform,, ether, | |
C1=CN=CC=C1C2=CC=NC=C2 | |
156.19 | |
CHEBI:30985 | |
Powder |
Chemical Identifiers
553-26-4 | |
156.19 | |
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridine, 4,4'-dipyridyl, 4,4-bipyridyl, 4,4-dipyridyl, 4-4-pyridyl pyridine, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb | |
CHEBI:30985 | |
C1=CN=CC=C1C2=CC=NC=C2 |
C10H8N2 | |
MWVTWFVJZLCBMC-UHFFFAOYSA-N | |
11107 | |
4-pyridin-4-ylpyridine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist
GHS Signal Word: Danger
EINECSNumber : 209-036-3