missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Dipyridyl, 98%, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC276200050

 View more versions of this product

Catalog No. AC276200050



Description

Description

Specifications

Specifications

4, 4'-Dipyridyl
Glass bottle
1% Max. (K.F.)
White to Beige
5g
98%
(C5H4N)-(C5H4N)
16,25
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridine, 4,4'-dipyridyl, 4,4-bipyridyl, 4,4-dipyridyl, 4-4-pyridyl pyridine, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4-pyridin-4-ylpyridine
11107
156.19
98%
Authentic
0.1% max.
305.0°C
110.0°C to 114.0°C
553-26-4
C10H8N2
23,2
15,3389
Solubility in water: 4,5g/L (25°C). Other solubilities: freely soluble in alcohol, benzene, chloroform,, ether,
C1=CN=CC=C1C2=CC=NC=C2
156.19
CHEBI:30985
Powder

Chemical Identifiers

553-26-4
156.19
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridine, 4,4'-dipyridyl, 4,4-bipyridyl, 4,4-dipyridyl, 4-4-pyridyl pyridine, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
CHEBI:30985
C1=CN=CC=C1C2=CC=NC=C2
C10H8N2
MWVTWFVJZLCBMC-UHFFFAOYSA-N
11107
4-pyridin-4-ylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist

GHS Signal Word: Danger

EINECSNumber : 209-036-3

SDS Documents