Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Diphenylbenzophenone, 97%

$306.31 - $306.31

Chemical Identifiers

Chemical Name or Material 4,4'-Diphenylbenzophenone
CAS 3478-90-8
Molecular Formula C25H18O
Formula Weight 334.42
MDL Number MFCD00016575
Beilstein 1885447
Synonym Di-4-biphenylyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0650214

  SDS
alfa aesar™
L0650214
25g Each for $306.31
Description & Specifications

Specifications

Formula Weight 334.42
Chemical Name or Material 4,4'-Diphenylbenzophenone
Molecular Formula C25H18O
MDL Number MFCD00016575
CAS 3478-90-8
Synonym Di-4-biphenylyl ketone
Assay 97%
Beilstein 1885447
Melting Point 235° to 240°C
Quantity 25g